Learn More
Cayman Chemical Tegaserod (maleate), Purity: ≥98%, CAS: 189188-57-6, 50 mg
SDP
Catalog No. 502970078
Encompass_Preferred
A potent agonist of the serotonin 4 receptor (5-HT4, Ki = 3.9-31 nM; also acts as an antagonist for the 5-HT2A, 5-HT2B, and 5-HT2C receptors (Ki = 32, 3.9, and 100 nM, respectively); stimulates colonic motility and transit in vivo
Catalog No. 50-297-0078 Supplier Cayman Chemical Supplier No. 1469250 MG
A potent agonist of the serotonin 4 receptor (5-HT4, Ki = 3.9-31 nM; also acts as an antagonist for the 5-HT2A, 5-HT2B, and 5-HT2C receptors (Ki = 32, 3.9, and 100 nM, respectively); stimulates colonic motility and transit in vivo