Learn More
Selleck Chemical LLC Tegatrabetan5mg
SDP
Supplier:  Selleck Chemical LLC S07335MG
Encompass_Preferred
Tegatrabetan (BC2059) is an antagonist of?-Catenin. Tegatrabetan (BC2059) treatment disrupts the binding of ?-catenin with the scaffold protein transducin ?-like 1 (TBL1) and proteasomal degradation and declines in the nuclear levels of ?-catenin. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3044