Learn More
Medchemexpress LLC HY-16482 1mg Medchemexpress, Teglicar CAS:250694-07-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY164821MG
Encompass_Preferred
Medchemexpress, HY-16482 1mg Teglicar CAS:250694-07-6 Teglicar is a selective and reversible inhibitor of liver isoform of carnitine palmitoyl-transferase 1 (L-CPT1). Teglicar reduces ketogenesis and glucose production, decreases gluconeogenesis and improves glucose homeostasis. Teglicar has a potential antihyperglycemic propert. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0718