Learn More
Medchemexpress LLC Teglicar | 250694-07-6 | 98.0% | 399.61 | 5 MG
SDP
Catalog No. 5000390998
Encompass_Preferred
Teglicar is a selective and reversible orally active liver isoform of carnitine palmitoyl-transferase 1 (L-CPT1) inhibitor with an IC50 value of 0.68 μM and a Ki value of 0.36 μM. Teglicar has a potential antihyperglycemic property. It can be used for the research of diabetes and neurodegenerative disease including Huntington's disease (HD).
  • Selective and reversible orally active liver isoform of carnitine palmitoyl-transferase 1 (L-CPT1) inhibitor.
  • Has an IC50 value of 0.68 μM and a Ki value of 0.36 μM for L-CPT1.
  • Possesses potential antihyperglycemic properties.
  • Suitable for research on diabetes and neurodegenerative diseases, such as Huntington's disease (HD).
Catalog No. 50-003-90998 Supplier Medchemexpress LLC Supplier No. HY164825MG
Teglicar is a selective and reversible orally active liver isoform of carnitine palmitoyl-transferase 1 (L-CPT1) inhibitor with an IC50 value of 0.68 μM and a Ki value of 0.36 μM. Teglicar has a potential antihyperglycemic property. It can be used for the research of diabetes and neurodegenerative disease including Huntington's disease (HD).
  • Selective and reversible orally active liver isoform of carnitine palmitoyl-transferase 1 (L-CPT1) inhibitor.
  • Has an IC50 value of 0.68 μM and a Ki value of 0.36 μM for L-CPT1.
  • Possesses potential antihyperglycemic properties.
  • Suitable for research on diabetes and neurodegenerative diseases, such as Huntington's disease (HD).