Learn More
Medchemexpress LLC HY-17623 5mg Medchemexpress, Tegoprazan CAS:942195-55-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY176235MG
Encompass_Preferred
Medchemexpress, HY-17623 5mg Tegoprazan CAS:942195-55-3 Tegoprazan, a potassium-competitive acid blocker, is a potent, oral active and highly selective inhibitor of gastric H+/K+-ATPase that could control gastric acid secretion and motility, with IC50 values ranging from 0.29-0.52 μM for porcine, canine, and human H+/K+-ATPases in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2003