Learn More
Medchemexpress LLC Tegoprazan | 942195-55-3 | 99.8% | 387.38 | 100 MG
SDP
Catalog No. 5000387477
Encompass_Preferred
Tegoprazan is a potassium-competitive acid blocker, a reversible, orally active, and highly selective inhibitor of gastric H+/K+-ATPase. It inhibits gastric acid secretion and motility against porcine, canine, and human H+/K+-ATPase with IC50 values ranging from 0.29-0.52 μM in vitro. It also significantly improves colitis and enhances the intestinal epithelial barrier function in mice, making it promising for research of inflammatory bowel, gastric acid-related, and motility-impaired diseases.
  • Reversible, orally active, and highly selective inhibitor of gastric H+/K+-ATPase.
  • Inhibits gastric acid secretion and motility.
  • Improves colitis and enhances intestinal epithelial barrier function.
  • Promising for research of inflammatory bowel, gastric acid-related, and motility-impaired diseases.
Catalog No. 50-003-87477 Supplier Medchemexpress LLC Supplier No. HY17623100MG
Tegoprazan is a potassium-competitive acid blocker, a reversible, orally active, and highly selective inhibitor of gastric H+/K+-ATPase. It inhibits gastric acid secretion and motility against porcine, canine, and human H+/K+-ATPase with IC50 values ranging from 0.29-0.52 μM in vitro. It also significantly improves colitis and enhances the intestinal epithelial barrier function in mice, making it promising for research of inflammatory bowel, gastric acid-related, and motility-impaired diseases.
  • Reversible, orally active, and highly selective inhibitor of gastric H+/K+-ATPase.
  • Inhibits gastric acid secretion and motility.
  • Improves colitis and enhances intestinal epithelial barrier function.
  • Promising for research of inflammatory bowel, gastric acid-related, and motility-impaired diseases.