Learn More
Medchemexpress LLC Tegoprazan | 942195-55-3 | 99.8% | 387.38 | 250 MG
SDP
Catalog No. 5000383915
Encompass_Preferred
Tegoprazan (CJ-12420) is a potassium-competitive acid blocker and a reversible, orally active, and highly selective inhibitor of gastric H+/K+-ATPase. It inhibits gastric acid secretion and motility in vitro. This compound improves colitis and enhances intestinal epithelial barrier function in mice, making it promising for research related to inflammatory bowel, gastric acid-related, and motility-impaired diseases.
  • Potassium-competitive acid blocker
  • Reversible, orally active
  • Highly selective inhibitor of gastric H+/K+-ATPase
  • Inhibits gastric acid secretion and motility
  • Improves colitis
  • Enhances intestinal epithelial barrier function
Catalog No. 50-003-83915 Supplier Medchemexpress LLC Supplier No. 250MGHY17623
Tegoprazan (CJ-12420) is a potassium-competitive acid blocker and a reversible, orally active, and highly selective inhibitor of gastric H+/K+-ATPase. It inhibits gastric acid secretion and motility in vitro. This compound improves colitis and enhances intestinal epithelial barrier function in mice, making it promising for research related to inflammatory bowel, gastric acid-related, and motility-impaired diseases.
  • Potassium-competitive acid blocker
  • Reversible, orally active
  • Highly selective inhibitor of gastric H+/K+-ATPase
  • Inhibits gastric acid secretion and motility
  • Improves colitis
  • Enhances intestinal epithelial barrier function