Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sonoco™ Tegrant ThermoSafe LabMailer Absorbent Sheets

Catalog No. p-4919710 Shop All Sonoco ThermoSafe Products
Change view
Click to view available options
Capacity:
22.5 mL of blood/sheet
360 mL of water/90 mL blood/sheet
Diameter (Metric) Interior:
15.2 x 15.2 cm
30.4 x 30.4 cm
7.6 x 7.6 cm
Diameter (English) Interior:
3 x 3 in.
6 x 6 in.
12 x 12 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Diameter (Metric) Interior Diameter (English) Interior
03-531-62 22.5 mL of blood/sheet 7.6 x 7.6 cm 3 x 3 in.
03-600-160 360 mL of water/90 mL blood/sheet 15.2 x 15.2 cm 6 x 6 in.
03-600-161 360 mL of water/90 mL blood/sheet 30.4 x 30.4 cm 12 x 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-531-62 Supplier Sonoco™ Supplier No. ZORB33
Only null left
Add to Cart
Add to Cart

Lightweight, economical lab mailer shippers

  • Accommodates VacuTainer™, Microtainer™, and other collection/transport tubes
  • Available in multi-tube, slide, and tube/slide combinations
  • Polystyrene foam construction saves postage
  • Reusable and recyclable

Additional packaging may be required to conform to CFR72.2 specification (See Packaging Accessories Kit Mfr. No. 471).

TRUSTED_SUSTAINABILITY

Specifications

Quantity 22.5mL of blood per sheet
For Use With (Equipment) Provide maximum breakage protection for known non-hazardous materials.
Applications For protection of frozen, refrigerated, and ambient specimens required for compliance, analysis, testing, research and development, public health, industrial hygiene and environmental use.
Diameter (English) Interior 3 x 3 in.
Diameter (Metric) Interior 7.6 x 7.6 cm
Capacity 22.5 mL of blood/sheet
Material Lightweight Polystyrene Foam
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.