Sonoco™ Tegrant ThermoSafe LabMailer Absorbent Sheets

Lightweight, economical lab mailer shippers

Manufacturer: Sonoco™ ZORB33

 View more versions of this product

Catalog No. 03-531-62

  Check Availability
Add to Cart

    Specifications

    Diameter (English) Interior 3 x 3 in.
    Diameter (Metric) Interior 7.6 x 7.6cm
    For Use With For protection of frozen, refrigerated, and ambient specimens required for compliance, analysis, testing, research and development, public health, industrial hygiene and environmental use.
    Capacity Holds 22.5mL of blood per sheet
    For Use With (Equipment) Provide maximum breakage protection for known non-hazardous materials.
    Material Lightweight polystyrene foam
    Quantity 22.5mL of blood per sheet

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit