Learn More
Medchemexpress LLC HY-12398 1mg Medchemexpress, TEI-9647 CAS:173388-20-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY123981MG
Encompass_Preferred
Medchemexpress, HY-12398 1mg TEI-9647 CAS:173388-20-0 TEI-9647 is a potent and specific vitamin D receptor (VDR) antagonist. TEI-9647, a Vitamin D3 Lactone analogue, inhibits bone resorption induced by of 1α,25-(OH)2D3. TEI-9647 has the potential for suppressing the excessive bone resorption and osteoclast formation in Paget's disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1031