Learn More
Medchemexpress LLC HY-12398A 1mg Medchemexpress, TEI-9648 CAS:173388-21-1 Purity:>98%
SDP
Catalog No. 502081033
Encompass_Preferred
Medchemexpress, HY-12398A 1mg TEI-9648 CAS:173388-21-1 TEI-9648 is a potent and specific vitamin D receptor (VDR) antagonist. TEI-9648, a Vitamin D3 Lactone analogue, inhibits bone resorption induced by of 1α,25-(OH)2D3. TEI-9648 has the potential for bone metabolism research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1033 Supplier Medchemexpress LLC Supplier No. HY12398A1MG
Add to Cart
Add to Cart
Medchemexpress, HY-12398A 1mg TEI-9648 CAS:173388-21-1 TEI-9648 is a potent and specific vitamin D receptor (VDR) antagonist. TEI-9648, a Vitamin D3 Lactone analogue, inhibits bone resorption induced by of 1α,25-(OH)2D3. TEI-9648 has the potential for bone metabolism research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.