Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Teicoplanin, G-Biosciences
SCI_ED

Catalog No. S139829
Change view
Click to view available options
Quantity:
100 mg
1000 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139829 1000 mg
S139787 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139829 Supplier G-Biosciences Supplier No. RC1114
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Teicoplanin is a glycopeptide antibiotic isolated from Actinoplanes teichomyceticus. It is a mixture of several teicoplanins.

Teicoplanin is effective against gram-positive bacteria resistant to Beta-lactam antibiotics. It has been effective against methicillin-resistant Staphylococcus aureus and Enterococcus faecalis.

Chemical Identifiers

Molecular Weight (g/mol) 1879.66

Specifications

Physical Form Powder
Quantity 1000 mg
Product Line Gibco
Percent Purity >95%
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.