Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Teicoplanin, Thermo Scientific™

Catalog No. AC455400010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455400010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC455400010 Supplier Thermo Scientific Chemicals Supplier No. 455400010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Chemical Identifiers

CAS 61036-62-2
Molecular Formula C88H97Cl2N9O33
Molecular Weight (g/mol) 1879.67
MDL Number MFCD05664625
InChI Key BJNLLBUOHPVGFT-QRZIFLFXSA-N
Synonym Targocid, Teichomycin
PubChem CID 133640186
IUPAC Name (1S,2R,19R,22R,34S,37R,40R,52R)-22-amino-63,65-dichloro-64-{[(2S,3R,4R,5S,6R)-3-decanamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2-{[(2R,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-26,31,44,49-tetrahydroxy-21,35,38,54,56,59-hexaoxo-47-{[(2R,3S,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-7,13,28-trioxa-20,36,39,53,55,58-hexaazaundecacyclo[38.14.2.2³,⁶.2¹⁴,¹⁷.2¹⁹,³⁴.1⁸,¹².1²³,²⁷.1²⁹,³³.1⁴¹,⁴⁵.0¹⁰,³⁷.0⁴⁶,⁵¹]hexahexaconta-3,5,8,10,12(64),14,16,23,25,27(61),29,31,33(60),41,43,45(57),46,48,50,62,65-henicosaene-52-carboxylic acid
SMILES CCCCCCCCCC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1OC1=C2OC3=CC=C(C[C@H]4NC(=O)[C@H](N)C5=CC=C(O)C(OC6=CC(O)=CC(=C6)[C@H](NC4=O)C(=O)N[C@@H]4C(C=C1OC1=CC=C(C=C1Cl)[C@@H](O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1NC(C)=O)[C@@H]1NC(=O)[C@H](NC4=O)C4=CC=C(O)C(=C4)C4=C(O[C@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]6O)C=C(O)C=C4[C@@H](NC1=O)C(O)=O)=C2)=C5)C=C3Cl

Specifications

Type Teicoplanin
Physical Form Powder
Melting Point 260.0°C
Color White to Yellow
Quantity 1 g
Chemical Name or Material Teicoplanin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.