Learn More
Cayman Chemical Teicoplanin A2-3, Purity: ≥85%, CAS: 91032-36-9, 5 mg
SDP
Catalog No. 502983533
Encompass_Preferred
A major component of the teicoplanin complex, a glycopeptide antibiotic produced by A. teichomyceticus that is broadly effective against Gram-positive bacteria
Catalog No. 50-298-3533 Supplier Cayman Chemical Supplier No. 201885 MG
A major component of the teicoplanin complex, a glycopeptide antibiotic produced by A. teichomyceticus that is broadly effective against Gram-positive bacteria