Learn More
Cayman Chemical Teicoplanin A2-5, Purity: ≥95%, CAS: 91032-38-1, 5 mg
SDP
Catalog No. 502981842
Encompass_Preferred
A non-polar glycopeptide antibiotic produced by A. teichomyceticus that is broadly effective against Gram-positive bacteria
Catalog No. 50-298-1842 Supplier Cayman Chemical Supplier No. 201925 MG
A non-polar glycopeptide antibiotic produced by A. teichomyceticus that is broadly effective against Gram-positive bacteria