Learn More
Cayman Chemical Teicoplanin Complex, Purity: ≥95%, CAS: 61036-62-2, 100 mg
SDP
Catalog No. 502969368
Encompass_Preferred
A mixture of closely related metabolites produced by A. teichomyceticus that are effective against Gram-positive bacteria; prevents bacterial cell-wall synthesis by binding and sequestering a cell-wall precursor with a D-alanine-containing peptide
Catalog No. 50-296-9368 Supplier Cayman Chemical Supplier No. 15511100 MG
A mixture of closely related metabolites produced by A. teichomyceticus that are effective against Gram-positive bacteria; prevents bacterial cell-wall synthesis by binding and sequestering a cell-wall precursor with a D-alanine-containing peptide