Learn More
Cayman Chemical Teijin Compound 1 (hydrochloride), Purity: ≥98%, CAS: 1313730-14-1, 10 mg
SDP
Catalog No. 502989505
Encompass_Preferred
A CCR2b antagonist (IC50 = 180 nM in a radioligand binding assay); inhibits CCL2-induced chemotaxis in THP-1 cells (EC50 = 24 nM)
Catalog No. 50-298-9505 Supplier Cayman Chemical Supplier No. 3097710 MG
A CCR2b antagonist (IC50 = 180 nM in a radioligand binding assay); inhibits CCL2-induced chemotaxis in THP-1 cells (EC50 = 24 nM)