Learn More
IMEB, Inc Tek-Select Cassette Filing Cabinet 6 Drawer, Stackable. Tek-Select Cassette Filing Cabinet provide a permanent storage reference unit for cassettes and rings.
SDP
Catalog No. NC2341077
Encompass_Preferred
Tek-Select Cassette Filing Cabinet provides a permanent storage reference unit for cassettes and rings. The filing cabinets are stack-able and accommodate 1,500 cassettes or 1,000 rings. One piece construction eliminates hinges and doors for dust-free storage. Includes 6 impact resistant plastic drawers with identification area writing surface. Dimensions: 15″L x 9″W x 17″H. (Compare to SAKURA 4192 file.)
Catalog No. NC2341077 Supplier IMEB, Inc Supplier No. 4192TS
Tek-Select Cassette Filing Cabinet provides a permanent storage reference unit for cassettes and rings. The filing cabinets are stack-able and accommodate 1,500 cassettes or 1,000 rings. One piece construction eliminates hinges and doors for dust-free storage. Includes 6 impact resistant plastic drawers with identification area writing surface. Dimensions: 15″L x 9″W x 17″H. (Compare to SAKURA 4192 file.)