Learn More
Selleck Chemical LLC Telacebec
SDP
Catalog No. 5000001398
Encompass_Preferred
Telacebec is a novel antituberculosis drug that targets Mycobacterium tuberculosis cellular energy production through inhibition of the mycobacterial cytochrome bc1 complex Telacebec (Q203 IAP6) is active against Mycobacterium tuberculosis H37Rv with MIC50 of 2 7 nM in culture broth medium
Catalog No. 50-000-01398 Supplier Selleck Chemical LLC Supplier No. S773925MG
Telacebec is a novel antituberculosis drug that targets Mycobacterium tuberculosis cellular energy production through inhibition of the mycobacterial cytochrome bc1 complex Telacebec (Q203 IAP6) is active against Mycobacterium tuberculosis H37Rv with MIC50 of 2 7 nM in culture broth medium