Learn More
TARGETMOL CHEMICALS INC TELAGLENASTAT 25MG
SDP
Catalog No. 502576451
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Telaglenastat (CB 839) (IC50 of 24 nM) an effective specific and oral inhibitor which is bioavailable glutaminase for recombinant human GAC. purity: 99%
Catalog No. 50-257-6451 Supplier TARGETMOL CHEMICALS INC Supplier No. T679725MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Telaglenastat (CB 839) (IC50 of 24 nM) an effective specific and oral inhibitor which is bioavailable glutaminase for recombinant human GAC. purity: 99%