Learn More
Selleck Chemical LLC Telaglenastat
Supplier: Selleck Chemical LLC S7655100MG
Telaglenastat (CB-839) is a potent selective and orally bioavailable glutaminase inhibitor with IC50 of 24 nM for recombinant human GAC CB-839(Telaglenastat) inudces autophagy and has antitumor activity Phase 1