Learn More
Selleck Chemical LLC Telaglenastat
SDP
Catalog No. 5000001536
Encompass_Preferred
Telaglenastat (CB-839) is a potent selective and orally bioavailable glutaminase inhibitor with IC50 of 24 nM for recombinant human GAC CB-839(Telaglenastat) inudces autophagy and has antitumor activity Phase 1
Catalog No. 50-000-01536 Supplier Selleck Chemical LLC Supplier No. S765525MG
Telaglenastat (CB-839) is a potent selective and orally bioavailable glutaminase inhibitor with IC50 of 24 nM for recombinant human GAC CB-839(Telaglenastat) inudces autophagy and has antitumor activity Phase 1