Learn More
TARGETMOL CHEMICALS INC TELAPREVIR 10MG
SDP
Catalog No. 502579495
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Telaprevir (Incivek) (VX-950) is an effective inhibitor of HCV NS3-4A serine protease (IC50 0.35 uM). purity: 96%
Catalog No. 50-257-9495 Supplier TARGETMOL CHEMICALS INC Supplier No. T619010MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Telaprevir (Incivek) (VX-950) is an effective inhibitor of HCV NS3-4A serine protease (IC50 0.35 uM). purity: 96%