Learn More
Medchemexpress LLC HY-10235 100mg , Telaprevir VX-950 CAS:402957-28-2 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY10235100MG
Encompass_Preferred
Medchemexpress, HY-10235 100mg Telaprevir VX-950 CAS:402957-28-2 Formula:C36H53N7O6 Ki: 7 nM (genotype 1 HCV NS3-4A protease) Purity:98% Telaprevir is a highly selective, reversible, and potent peptidomimetic inhibitor of the HCV NS3-4A protease , the steady-state inhibitory constant ( K i ) of Telaprevir is 7 nM against a genotype 1 (H strain) NS3 protease domain plus a NS4A cofactor peptide. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3284
 Restricted Item