Learn More
Cayman Chemical TelaprevIr-d4 250ug
SDP
Catalog No. 502991118
Encompass_Preferred
An internal standard for the quantification of telaprevir by GC- or LC-MS
Catalog No. 50-299-1118 Supplier Cayman Chemical Supplier No. 31317250 UG
An internal standard for the quantification of telaprevir by GC- or LC-MS