Learn More
Apexbio Technology LLC Telaprevir (VX-950), 402957-28-2, MFCD11616089, 10 mg
SDP
Supplier:  Apexbio Technology LLC A403110
Encompass_Preferred
MF: C36H53N7O6, MW: 679.9, Solubility: 32.95 mg/mL in DMSO,insoluble in EtOH,insoluble in H2O. Telaprevir (also known as VX-950), derived from the viral NS5A/5B substrate of the protease through structure-based techniques, is a novel and potent inhibitor of hepatitis C virus (HCV) NS3-4A protease, which covalently and reversibly inhibit the NS3-4A protease via a slow-binding and slow-dissociation mechanism. Results of multiple studies show that telaprevir potently inhibits a genotype 1 (H strain) NS3 protease domain plus a NS4A cofactor peptide with the inhibitory constant (Ki) of 7 nM, exhibits strong anti-HCV in HCV replicon cells in vitro, and suppresses HCV replication as well as the emergence of resistance in HCV replicon cells with a synergistic effect with IFN-alpha.
Catalog No. 50-101-3645