Learn More
Selleck Chemical LLC Telaprevir (VX-950) 5mg 402957-28-2 LY-570310, MP-424
SDP
Catalog No. 507217
Encompass_Preferred
Telaprevir (VX-950, LY-570310, MP-424) is an HCV NS3-4A serine protease inhibitor with IC50 of 0.35 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-721-7 Supplier Selleck Chemical LLC Supplier No. S15385MG
Add to Cart
Add to Cart
Telaprevir (VX-950, LY-570310, MP-424) is an HCV NS3-4A serine protease inhibitor with IC50 of 0.35 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.