Learn More
Selleck Chemical LLC Telaprevir (VX-950) 50mg 402957-28-2 LY-570310, MP-424
SDP
Catalog No. 507220
Encompass_Preferred
Telaprevir (VX-950, LY-570310, MP-424) is an HCV NS3-4A serine protease inhibitor with IC50 of 0.35 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-722-0 Supplier Selleck Chemical LLC Supplier No. S153850MG
Telaprevir (VX-950, LY-570310, MP-424) is an HCV NS3-4A serine protease inhibitor with IC50 of 0.35 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.