Learn More
Medchemexpress LLC HY-10527 5mg Medchemexpress, Telatinib CAS:332012-40-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY105275MG
Encompass_Preferred
Medchemexpress, HY-10527 5mg Telatinib CAS:332012-40-5 Telatinib (Bay 57-9352) is an orally active, small molecule inhibitor of VEGFR2, VEGFR3, PDGFα, and c-Kit with IC50s of 6, 4, 15 and 1 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8262