Learn More
Selleck Chemical LLC Telatinib 50mg 332012-40-5 BAY 57-9352
SDP
Supplier:  Selleck Chemical LLC S223150MG
Encompass_Preferred
Telatinib (BAY 57-9352) is a potent inhibitor of VEGFR2/3, c-Kit and PDGFR? with IC50 of 6 nM/4 nM, 1 nM and 15 nM, respectively. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-525-4