Learn More
Medchemexpress LLC Telavancin hydrochloride | 560130-42-9 | ≥98.0% | 1792.10 | 1 MG
SDP
Catalog No. 5000392142
Encompass_Preferred
Telavancin hydrochloride is a semisynthetic derivative of vancomycin that demonstrates antibacterial efficacy by inhibiting cell wall synthesis and disrupting cell membrane barrier function. It is effective against antibiotic-resistant Staphylococcus aureus strains, including MRSA, VISA, and hVISA. This product mitigates gram-positive bacteria-induced complicated skin and skin structure infections (cSSSI) and is intended for research use only.
  • Inhibits cell wall synthesis.
  • Disrupts cell membrane barrier function.
  • Effective against antibiotic-resistant Staphylococcus aureus strains (MRSA, VISA, hVISA).
  • Mitigates gram-positive bacteria-induced complicated skin and skin structure infections.
  • Intended for research use only.
Catalog No. 50-003-92142 Supplier Medchemexpress LLC Supplier No. HY164851MG
Telavancin hydrochloride is a semisynthetic derivative of vancomycin that demonstrates antibacterial efficacy by inhibiting cell wall synthesis and disrupting cell membrane barrier function. It is effective against antibiotic-resistant Staphylococcus aureus strains, including MRSA, VISA, and hVISA. This product mitigates gram-positive bacteria-induced complicated skin and skin structure infections (cSSSI) and is intended for research use only.
  • Inhibits cell wall synthesis.
  • Disrupts cell membrane barrier function.
  • Effective against antibiotic-resistant Staphylococcus aureus strains (MRSA, VISA, hVISA).
  • Mitigates gram-positive bacteria-induced complicated skin and skin structure infections.
  • Intended for research use only.