Learn More
Medchemexpress LLC HY-B0017 10mg , Telbivudine CAS:3424-98-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB001710MG
Encompass_Preferred
HY-B0017 10mg Telbivudine CAS: 3424-98-4 Telbivudine (Epavudine), an orally active thymidine nucleoside analog, is a potent antiviral inhibitor of hepatitis B virus (HBV) replication. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1113