Learn More
TARGETMOL CHEMICALS INC TELBIVUDINE 25MG
SDP
Catalog No. 502577364
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Telbivudine (NV 02B) is a Hepatitis B Virus Nucleoside Analog Reverse Transcriptase Inhibitor. The mechanism of action of telbivudine is as a Nucleoside Reverse Transcriptase Inhibitor. The chemical classification of telbivudine is Nucleoside Analog. purity: 99%
Catalog No. 50-257-7364 Supplier TARGETMOL CHEMICALS INC Supplier No. T151325MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Telbivudine (NV 02B) is a Hepatitis B Virus Nucleoside Analog Reverse Transcriptase Inhibitor. The mechanism of action of telbivudine is as a Nucleoside Reverse Transcriptase Inhibitor. The chemical classification of telbivudine is Nucleoside Analog. purity: 99%