Learn More
Cole-Parmer Megger CHM2000 Model Cable Height Meter (35ft. max height)
Catalog No. 50993474 Shop All Cole-Parmer Products
Encompass
These cable height meters utilize ultrasonic sound to determine the height of up to six overhead cables and wires. The instruments are designed to address specific measuring requirements on cables up to 75 feed (23 m). The BD121136 and BD121138 are designed primarily for power cable applications, while the BD121130 is designed for telecommunication applications.
The Cable Height Meters are capable of measuring the lowest or highest cables (provided it is within range). Additionally the spacing between the next five cables, by selection of "Bottom UP" mode or "Top Down" mode, can be displayed. Applications include measuring cable heights of: telephone lines, distribution lines, transmission lines, cable television installers and contractors), street lights, SAG (lowest point to grou
Catalog No. 50-993-474 Supplier Cole-Parmer Supplier No. 2001081
These cable height meters utilize ultrasonic sound to determine the height of up to six overhead cables and wires. The instruments are designed to address specific measuring requirements on cables up to 75 feed (23 m). The BD121136 and BD121138 are designed primarily for power cable applications, while the BD121130 is designed for telecommunication applications.
The Cable Height Meters are capable of measuring the lowest or highest cables (provided it is within range). Additionally the spacing between the next five cables, by selection of "Bottom UP" mode or "Top Down" mode, can be displayed. Applications include measuring cable heights of: telephone lines, distribution lines, transmission lines, cable television installers and contractors), street lights, SAG (lowest point to grou