Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Telenzepine dihydrochloride | 147416-96-4 | MFCD01529948 | 99.6% | 443.39 | C19H24Cl2N4O2S | 10 MG
SDP

Catalog No. 5000297282
Encompass_Preferred

Telenzepine dihydrochloride is a selective, orally active muscarinic M1 receptor antagonist supplied as a solid for research use. It exhibits high purity and is offered in milligram-scale pack sizes for laboratory experiments.

  • Selective M1 muscarinic receptor antagonist.
  • High purity (99.6%).
  • Molecular weight 443.39 and formula C19H24Cl2N4O2S.
  • Water solubility: 62.5 mg/mL with ultrasonic warming.
  • Recommended storage: sealed, away from moisture; solid at -20°C.
  • Available in multiple milligram pack sizes for research applications.

Catalog No. 50-002-97282 Supplier Medchemexpress LLC Supplier No. HYB1789A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Telenzepine dihydrochloride is a selective, orally active muscarinic M1 receptor antagonist supplied as a solid for research use. It exhibits high purity and is offered in milligram-scale pack sizes for laboratory experiments.

  • Selective M1 muscarinic receptor antagonist.
  • High purity (99.6%).
  • Molecular weight 443.39 and formula C19H24Cl2N4O2S.
  • Water solubility: 62.5 mg/mL with ultrasonic warming.
  • Recommended storage: sealed, away from moisture; solid at -20°C.
  • Available in multiple milligram pack sizes for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.