Learn More
MSC Wontide Telescoping Lift Master Boom, Structural Steel, 4000 Lb. Capacity, 80-144 in. Length, 1 ea.
Catalog No. 5000516558
Encompass
This telescoping lift master boom provides a versatile solution for converting forklifts into overhead lifting cranes. Fabricated from structural steel and robotically welded to meet ISO standards, this attachment ensures rugged durability and long-term service in demanding industrial environments.
  • Features an infinitely adjustable locking screw for precise positioning
  • Includes safety strap for secure attachment to forklift forks
  • Designed with clear capacity and operational instruction stickers for safety
  • Constructed from high-strength structural steel
  • Provides a load capacity of 4,000 lb.
  • Telescoping length adjusts from 80 in. to 144 in.
Catalog No. 50-005-16558 Supplier MSC Supplier No. 47345871
This telescoping lift master boom provides a versatile solution for converting forklifts into overhead lifting cranes. Fabricated from structural steel and robotically welded to meet ISO standards, this attachment ensures rugged durability and long-term service in demanding industrial environments.
  • Features an infinitely adjustable locking screw for precise positioning
  • Includes safety strap for secure attachment to forklift forks
  • Designed with clear capacity and operational instruction stickers for safety
  • Constructed from high-strength structural steel
  • Provides a load capacity of 4,000 lb.
  • Telescoping length adjusts from 80 in. to 144 in.