Learn More
MSC PEC Tools, 3-1/2"-6" TELESCOPE GAGE
Catalog No. 5000513468
Encompass
The telescope gage is a precision measuring tool designed for accurately determining the internal dimensions of bores, slots, and recesses. It provides reliable measurements when used in conjunction with a micrometer, allowing for the indirect measurement of difficult-to-reach internal features.
  • Provides precision measurement of internal diameters
  • Ideal for use in bores, slots, and recesses
  • Compatible with micrometers for accurate dimension verification
  • Features a rugged construction for long-term durability
Catalog No. 50-005-13468 Supplier MSC Supplier No. 01471168
The telescope gage is a precision measuring tool designed for accurately determining the internal dimensions of bores, slots, and recesses. It provides reliable measurements when used in conjunction with a micrometer, allowing for the indirect measurement of difficult-to-reach internal features.
  • Provides precision measurement of internal diameters
  • Ideal for use in bores, slots, and recesses
  • Compatible with micrometers for accurate dimension verification
  • Features a rugged construction for long-term durability