Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional™ Telescopic Handle, Maxi

Catalog No. 19200817 Shop All Vileda Professional Products
Change view
Click to view available options
Autoclavable:
Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
19-200-817 Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-200-817 Supplier Vileda Professional™ Supplier No. 162238
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specialty above-the-floor tools are ideal for surfaces that are difficult to reach, clean and disinfect. Tools are easy to use and ergonomic allowing full range of motion during cleaning and disinfecting.

  • Aluminum handle extends from 9 ft. (274 cm) to 15 ft. (457.2 cm)
  • Lightweight, ergonomic and extrememly durable
  • Autoclavable
  • Color coded rings (blue, red, yellow green) to reduce cross contamination
  • Can be used with all CE Above The Floor Tools™

Specifications

Product Type Frame Handle
Color Silver, Blue
Sterility Non-sterile
Autoclavable Autoclavable
Size Extends from 9 to 15 feet
Quantity 5
For Use With (Application) All CE Above The Floor Tools
Material Aluminum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.