Learn More
BURKLE INC Telescopic rod, infinitely adj. 45.28–118.11 in. (115–300 cm), 3 rods
Catalog No. NC2187709
Encompass_Preferred
Different tools can be attached to the telescopic rod (snap-in joint). The individual rods have been provided with an end stop. This means that one rod cannot accidentally slip out of another rod.
The telescopic rods cannot be rotated radially, which means even better handling!
Telescopic rod 5355-0280, extremely short model – yet extendable up to 280 cm (110.24 in.). Fits into every car trunk!
This item is not returnable. View return policy
Catalog No. NC2187709 Supplier BURKLE INC Supplier No. 53550300
Different tools can be attached to the telescopic rod (snap-in joint). The individual rods have been provided with an end stop. This means that one rod cannot accidentally slip out of another rod.
The telescopic rods cannot be rotated radially, which means even better handling!
Telescopic rod 5355-0280, extremely short model – yet extendable up to 280 cm (110.24 in.). Fits into every car trunk!