Learn More
BURKLE INC Telescopic rod, infinitely adj. 64.96–177.17 in. (165–450 cm), 3 rods
Catalog No. 5000025909
Encompass_Preferred
Different tools can be attached to the telescopic rod (snap-in joint). The individual rods have been provided with an end stop. This means that one rod cannot accidentally slip out of another rod.
The telescopic rods cannot be rotated radially, which means even better handling!
Telescopic rod 5355-0280, extremely short model – yet extendable up to 280 cm (110.24 in.). Fits into every car trunk!
Catalog No. 50-000-25909 Supplier BURKLE INC Supplier No. 53550450
Different tools can be attached to the telescopic rod (snap-in joint). The individual rods have been provided with an end stop. This means that one rod cannot accidentally slip out of another rod.
The telescopic rods cannot be rotated radially, which means even better handling!
Telescopic rod 5355-0280, extremely short model – yet extendable up to 280 cm (110.24 in.). Fits into every car trunk!