Learn More
Research Products International Corp Polypropylene Micro-Tube Storage Boxes for 0.5 ml Tubes, 50 Tube Capacity, Natural, 12 per Case
SDP
Catalog No. 50212372
Encompass
Micro-tube storage boxes are constructed of heavy-duty polypropylene that can be autoclaved. Boxes are reusable and will not saturate or deteriorate like cardboard boxes. Translucent plastic lid allows visual sample identification. Boxes have a telescoping lid with a 50 tube capacity.
Catalog No. 50-212-372 Supplier Research Products International Corp Supplier No. 147905
Micro-tube storage boxes are constructed of heavy-duty polypropylene that can be autoclaved. Boxes are reusable and will not saturate or deteriorate like cardboard boxes. Translucent plastic lid allows visual sample identification. Boxes have a telescoping lid with a 50 tube capacity.