Learn More
Cole-Parmer Burkle 5355-0600 Telescoping rod, aluminum, 65; 120 cm; 4 rods
Catalog No. NC2171939 Shop All Cole-Parmer Products
Encompass_Preferred
This modular sampling system allows you to build a sampler according to your application. Select a telescoping aluminum rod with the length you need, then choose one or more beakers and/or a bottle holder to attach to the rod. Angled and pendulum beakers are available in plastic or metal, and the bottle holder fits plastic, glass, or metal bottles.
This item is not returnable. View return policy
Catalog No. NC2171939 Supplier Cole-Parmer Supplier No. 0604215
This modular sampling system allows you to build a sampler according to your application. Select a telescoping aluminum rod with the length you need, then choose one or more beakers and/or a bottle holder to attach to the rod. Angled and pendulum beakers are available in plastic or metal, and the bottle holder fits plastic, glass, or metal bottles.