Learn More
MSC Milwaukee Tool Pole Saw Chain, 15 in, 1 ea
Catalog No. 502869150
Encompass
This pole saw chain is designed for use with the Milwaukee M18 FUEL Telescoping Pole Saw. It offers high-quality performance and durability for demanding cutting applications.
  • Blued cutters deliver high-quality protection against corrosion
  • High-strength alloy steel provides greater durability
  • Low kickback saw chain
  • For use with narrow kerf bars
  • Pitch: .325 low profile
  • Gauge: .043 in
  • Includes 46 drive links
  • File size: 5/32 in
  • File angle: 30 degrees
Catalog No. 50-286-9150 Supplier MSC Supplier No. 05775382
This pole saw chain is designed for use with the Milwaukee M18 FUEL Telescoping Pole Saw. It offers high-quality performance and durability for demanding cutting applications.
  • Blued cutters deliver high-quality protection against corrosion
  • High-strength alloy steel provides greater durability
  • Low kickback saw chain
  • For use with narrow kerf bars
  • Pitch: .325 low profile
  • Gauge: .043 in
  • Includes 46 drive links
  • File size: 5/32 in
  • File angle: 30 degrees