Learn More
Selleck Chemical LLC Telisotuzumab vedotin-1MG-5
SDP
Catalog No. 503134765
Encompass_Preferred
Telisotuzumab vedotin (Teliso-V ABBV-399) is an antibody-drug conjugate (ADC) composed of the anti c-Met humanized monoclonal antibody coupled to the cytotoxic monomethyl auristatin E (MMAE) through a mc-val-cit-PABC type linker Telisotuzumab vedotin can be used for research on advanced solid tumours
Catalog No. 50-313-4765 Supplier Selleck Chemical LLC Supplier No. D40461MG5
Telisotuzumab vedotin (Teliso-V ABBV-399) is an antibody-drug conjugate (ADC) composed of the anti c-Met humanized monoclonal antibody coupled to the cytotoxic monomethyl auristatin E (MMAE) through a mc-val-cit-PABC type linker Telisotuzumab vedotin can be used for research on advanced solid tumours