Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tellurium ICP Standard, 10,000 ppm Te in 10% HNO3, Ricca Chemical
SDP

Catalog No. PTE10KN100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PTE10KN100 100 mL Natural LDPE Bottle
PTE10KN500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PTE10KN100 Supplier Ricca Chemical Company Supplier No. PTE10KN100
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13494-80-9 , 7697-37-2 , 7732-18-5
Molecular Formula Te
Molecular Weight (g/mol) 127.60
MDL Number MFCD00134062
InChI Key PORWMNRCUJJQNO-UHFFFAOYSA-N
PubChem CID 6327182
ChEBI CHEBI:30452
IUPAC Name tellurium
SMILES [Te]

Specifications

Packaging Natural LDPE Bottle
CAS Min % 67.0, 30.24, 0.67
CAS Max % 70.0, 31.48, 0.7
Physical Form Liquid
Color Colorless
pH < 2
Type Standard
Quantity 100 mL
Concentration or Composition (by Analyte or Components) 1mL = 10mg Te (10, 000ppm Te)Te in 10% HNO3
Chemical Name or Material Tellurium ICP Standard
ShelfLife 18 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.