Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tellurium(IV) bromide, ultra dry, 99.999% (metals basis)

Catalog No. AA4343909
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4343909 10 g
AA4343918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4343909 Supplier Thermo Scientific Chemicals Supplier No. 04343909
Add to Cart

CAS: 10031-27-3 | Br4Te | 447.22 g/mol

Tellurium tetrabromide it acts as a chemical and organic intermediate. It is also acts a conductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-27-3
Molecular Formula Br4Te
Molecular Weight (g/mol) 447.22
MDL Number MFCD00049575
InChI Key PTYIPBNVDTYPIO-UHFFFAOYSA-N
Synonym tellurium tetrabromide, tellurium iv bromide, tellurium bromide tebr4 , t-4, unii-7a29efj1af, 7a29efj1af, tetrabromo-, e4-tellane, tebr4, acmc-1bozh, tellurium iv bromide, ultra dry
PubChem CID 82311
IUPAC Name tetrabromo-λ⁴-tellane
SMILES Br[Te](Br)(Br)Br

Specifications

Boiling Point 420°C (decomposition)
Melting Point 380°C
Physical Form Powder
Quantity 10 g
UN Number UN2923
Merck Index 14,9125
Solubility Information It reacts with water and ethanol.
Formula Weight 447.27
Percent Purity 99.999%
Packaging Ampule
Sensitivity Moisture sensitive
Density 4.31 g/mL at 15°C
Assay (metals basis)
Chemical Name or Material Tellurium (IV) bromide, Ultra dry
Show More Show Less
Hazard Category H301+H311-H314-H330-H335
Hazard Statement GHS H Statement
H300-H314-H318
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 233-090-7
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.