Learn More
Tellurium(IV) bromide, ultra dry, 99.999% (metals basis)
Description
Tellurium tetrabromide it acts as a chemical and organic intermediate. It is also acts a conductor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Specifications
| Boiling Point | 420°C (decomposition) |
| Melting Point | 380°C |
| Physical Form | Powder |
| Quantity | 50 g |
| Packaging | Ampule |
| Sensitivity | Moisture sensitive |
| Density | 4.31 g/mL at 15°C |
| UN Number | UN2923 |
| Merck Index | 14,9125 |
| Solubility Information | It reacts with water and ethanol. |
| Show More |
Safety and Handling
| Hazard Category | H301+H311-H314-H330-H335 |
| Hazard Statement | GHS H Statement H300-H314-H318 Fatal if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S. |
| EINECSNumber | 233-090-7 |
| TSCA | Yes |
| storageNote1 |
|
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.