Alfa Aesar™ Tellurium(IV) iodide, 99% (metals basis)

Manufacturer: Alfa Aesar™ 8779506

Quantity 5g

 View more versions of this product

Catalog No. AA8779506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7790-48-9
    Molecular Formula TeI4
    Formula Weight 635.22
    MDL Number MFCD00049578
    UN Number UN2923
    Merck Index 14,9127
    Synonym Tellurium tetraiodide
    Chemical Name or Material Tellurium (IV) iodide

    Specifications

    Formula Weight 635.22
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Acrid
    UN Number UN2923
    Merck Index 14,9127
    Quantity 5g
    Solubility Information Decomposes slowly in cold water to TeO2 and HI
    Health Hazard 1 Danger
    Sensitivity Moisture Sensitive
    Synonym Tellurium tetraiodide
    Melting Point 280°C
    Health Hazard 2 GHS H Statement
    H301-H314-H318
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Density 5.403g/mL at 15°C
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
    TSCA TSCA
    Physical Form Crystalline
    MDL Number MFCD00049578
    CAS 7790-48-9
    Molecular Formula TeI4
    Chemical Name or Material Tellurium (IV) iodide
    Assay 99%, (metals basis)

    GHS H Statement
    H301-H314-H318
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit