Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tellurium(IV) isopropoxide, 99.9% (metals basis), Thermo Scientific™

Catalog No. AA3656502
Change view
Click to view available options
0.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AA3656502 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3656502 Supplier Thermo Scientific Chemicals Supplier No. 3656502

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1795-64-8
Boiling Point 76°C (0.5mmHg)
Odor Alcohol-like
Molecular Formula C12H28O4Te
Quantity 0.5g
MDL Number MFCD00145354
UN Number UN1993
Sensitivity Light and moisture sensitive
Synonym tellurium iv isopropoxide, tetraisopropoxytellurium iv, tellurium iv isopropoxide metals basis
InChI Key PIJBAYXHEGHMNO-UHFFFAOYSA-N
SMILES CC(C)O[Te](OC(C)C)(OC(C)C)OC(C)C
IUPAC Name 2-{[tris(propan-2-yloxy)-λ⁴-tellanyl]oxy}propane
Molecular Weight (g/mol) 363.95
PubChem CID 15771447
Formula Weight 363.73
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Tellurium(IV) isopropoxide
Show More Show Less
Hazard Category H226-H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H226-H302-H312-H332-H315-H319
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.