Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tellurium(IV) oxide, tech.

Catalog No. AA1060818
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1060818 50 g
AA1060830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1060818 Supplier Thermo Scientific Chemicals Supplier No. 01060818
Add to Cart

CAS: 7446-07-3 | O2Te | 159.60 g/mol

It is used as an acousto-optic material and a conditional glass former. The glasses of tellurium oxide have high refractive indices. Owing to the transmission into mid-infrared part of the electromagnetic spectrum, tellurium oxide is of technological interest for optical waveguides. It is also employed in optical fibre amplification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-07-3
Molecular Formula O2Te
Molecular Weight (g/mol) 159.60
MDL Number MFCD00011263
InChI Key LAJZODKXOMJMPK-UHFFFAOYSA-N
Synonym tellurium iv oxide, tellurium oxide teo2, tellurite teo2, teo2, unii-397e9rke83, oxotellane oxide, telluride oxide, 6-chlorobutyric acid, acmc-1bglq
PubChem CID 62638
SMILES O=[Te]=O

Specifications

Melting Point 733°C
Physical Form Powder
Quantity 50 g
Solubility Information Practically insoluble in water. Soluble in NaOH solutions and HCl
IUPAC Name tellurium dioxide
Formula Weight 159.6
Grade Technical
Density 5.8
Chemical Name or Material Tellurium(IV) Oxide
Hazard Category H317-H332-H360
Precautionary Statement P201-P202-P261-P271-P272-P280g-P281-P302+P352-P304+P340-P308+P313-P333+P313-P363-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 231-193-1
RTECSNumber WY2675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.